Ethylenediaminetetraacetic dianhydride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

E0480-25G E0480-100G
TCE0480-025GEA 229.1 CAD
TCE0480-025G TCE0480-100G
Ethylenediaminetetraacetic dianhydride ≥98.0% (by titrimetric analysis)
Ethylenediaminetetraacetic dianhydride

CAS Number: 23911-25-3
MDL Number: MFCD00074963
Molecular Formula: C10H12N2O6
Molecular Weight: 256.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 190


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Analytical Science, Chelating Agents for Magnetic Resonance Imaging (MRI), Complexones (Chelating Agents)
Other Product Names / Synonyms: 4,4'-Ethylenebis(2,6-morpholinedione), EDTA Dianhydride

Formula: C₁₀H₁₂N₂O₆
Melting Pt: 190 °C
MDL Number: MFCD00074963
CAS Number: 23911-25-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=616d60c7f3e0bfd9e1adcb5c3c50adbecdd32797&vid=616d60c7f3e0bfd9e1adcb5c3c50adbecdd32797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR