3-Ethyl-2-methylbenzothiazolium iodide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

E0476-5G E0476-25G
TCE0476-5GEA 137.46 CAD
TCE0476-5G TCE0476-025G
3-Ethyl-2-methylbenzothiazolium iodide ≥98.0% (by HPLC, titration analysis)
3-Ethyl-2-methylbenzothiazolium iodide

CAS Number: 3119-93-5
MDL Number: MFCD00044134
Molecular Formula: C10H12INS
Molecular Weight: 305.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 196

Formula: C₁₀H₁₂INS
Melting Pt: 196 °C
MDL Number: MFCD00044134
CAS Number: 3119-93-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf5c9bfa6f8824e2cc0bad08911940dacbcfadb4&vid=bf5c9bfa6f8824e2cc0bad08911940dacbcfadb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR