Ethyl isopropyl sulfone ≥97.0%

Supplier: TCI America

E0475-25G E0475-5G
TCE0475-025GEA 235.42 CAD
TCE0475-025G TCE0475-5G
Ethyl isopropyl sulfone ≥97.0%
Ethyl isopropyl sulfone

CAS Number: 4853-75-2
MDL Number: MFCD00050418
Molecular Formula: C5H12O2S
Molecular Weight: 136.21
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 91
Specific Gravity (20/20): 1.09

Formula: C₅H₁₂O₂S
Boiling Pt: 91 °C
Density: 1.09 (20 °C)
MDL Number: MFCD00050418
CAS Number: 4853-75-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8d881d77e81e3da7796f8b923f773b9dfb87258&vid=c8d881d77e81e3da7796f8b923f773b9dfb87258&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR