Ethyl methyl sulfone ≥98.0%

Supplier: TCI America

Synonyms: Ethylmethylsulphone, Methylsulphonylethane, Ethylmethylsulfone, Methylsulfonylethane

E0474-25G E0474-100G
TCE0474-025GEA 344.44 CAD
TCE0474-025G TCE0474-100G
Ethyl methyl sulfone ≥98.0%
Ethyl methyl sulfone

CAS Number: 594-43-4
MDL Number: MFCD00015543
Molecular Formula: C3H8O2S
Molecular Weight: 108.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Freezing point (°C): 34


Other Product Names / Synonyms: Methylsulfonylethane

Formula: C₃H₈O₂S
MW: 108.16 g/mol
MDL Number: MFCD00015543
CAS Number: 594-43-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45e88e0097fb07add5e0dbaa3ab26110b986538d&vid=45e88e0097fb07add5e0dbaa3ab26110b986538d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR