4-Ethylamino-1-butanol ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
4-(Ethylamino)butan-1-ol, N-Ethylbutanolamine
E0473-25G
E0473-5G
TCE0473-025GEA
357.08
CAD
TCE0473-025G
TCE0473-5G
4-Ethylamino-1-butanol ≥98.0% (by titrimetric analysis)
4-Ethylamino-1-butanol
CAS Number: 39216-86-9
MDL Number: MFCD00671534
Molecular Formula: C6H15NO
Molecular Weight: 117.19
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 86
Specific Gravity (20/20): 0.91
Other Product Names / Synonyms: N-Ethylbutanolamine
Formula:
C₆H₁₅NO MW: 117.19 g/mol Boiling Pt: 86 °C Density: 0.91 (20 °C) |
MDL Number:
MFCD00671534 CAS Number: 39216-86-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...