Ethylene di(thiotosylate) ≥98.0%

Supplier: TCI America

Synonyms: 1,2-Ethanedithiol Ditosylate, S,S'-Ethylene p-Toluenethiosulfonate, Ethylene Bis(p-toluenethiosulphonate), S,S'-Ethylene p-Toluenethiosulphonate, Ethylene Bis(p-toluenethiosulfonate), 1,2-Di(p-tosylthio)ethane

E0471-5G E0471-25G
TCE0471-005GEA 317.42 CAD
TCE0471-005G TCE0471-025G
Ethylene di(thiotosylate) ≥98.0%
Ethylene di(thiotosylate)

[Protecting Reagent for Active Methylene]
CAS Number: 2225-23-2
MDL Number: MFCD00008550
Molecular Formula: C16H18O4S4
Molecular Weight: 402.56
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 74


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: 1,2-Di(p-tosylthio)ethane, 1,2-Ethanedithiol Ditosylate, Ethylene Bis(p-toluenethiosulfonate), S,S'-Ethylene p-Toluenethiosulfonate

Formula: C₁₆H₁₈O₄S₄
MW: 402.56 g/mol
Melting Pt: 74 °C
MDL Number: MFCD00008550
CAS Number: 2225-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94e044356e47e810b5bb61b2c6fadc4e73890add&vid=94e044356e47e810b5bb61b2c6fadc4e73890add&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR