Ethyl (R)-(+)-1-ethyl-2-pyrrolidinecarboxylate 99,0

Supplier: TCI America

Synonyms: (R)-(+)-1-Ethyl-2-pyrrolidinecarboxylic Acid Ethyl Ester, (+)-1-Ethyl-D-proline Ethyl Ester, Et-D-Pro-OEt

E0469-5G E0469-1G
TCE0469-5GEA 294.2 CAD
TCE0469-5G TCE0469-001G
Ethyl (R)-(+)-1-ethyl-2-pyrrolidinecarboxylate 99,0
Ethyl (R)-(+)-1-ethyl-2-pyrrolidinecarboxylate

CAS Number: 381670-33-3
MDL Number: MFCD00671533
Molecular Formula: C9H17NO2
Molecular Weight: 171.24
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 0.97
Specific rotation [a]20/D: 94 deg (C=0.6, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: (R)-(+)-1-Ethyl-2-pyrrolidinecarboxylic Acid Ethyl Ester, (+)-1-Ethyl-D-proline Ethyl Ester, Et-D-Pro-OEt

Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
Density: 0.97 (20 °C)
MDL Number: MFCD00671533
CAS Number: 381670-33-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0c6144f10d836d40e3bd95a29b1fb6d2d5ab345&vid=c0c6144f10d836d40e3bd95a29b1fb6d2d5ab345&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR