(R)-(+)-1-Ethyl-2-pyrrolidinecarboxamide

Supplier: TCI America

Synonyms: (R)-(+)-1-Ethyl-2-pyrrolidinecarboxamide, (R)-1-Ethylpyrrolidine-2-carboxamide, N-Ethyl-D-prolinamide

E0468-5G E0468-1G
TCE0468-5GEA 740.55 CAD
TCE0468-5G TCE0468-001G
(R)-(+)-1-Ethyl-2-pyrrolidinecarboxamide
(R)-(+)-1-Ethyl-2-pyrrolidinecarboxamide

CAS Number: 381670-32-2
MDL Number: MFCD00671532
Molecular Formula: C7H14N2O
Molecular Weight: 142.20
Form: Crystal
Color: White
Melting point (°C): 111
Specific rotation [a]20/D: 117 deg (C=0.5, MeOH)


Other Product Names / Synonyms: N-Ethyl-D-prolinamide

Formula: C₇H₁₄N₂O
Melting Pt: 111 °C
MDL Number: MFCD00671532
CAS Number: 381670-32-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7fb6546709e8ec395496e82415fab714cab0ac6&vid=f7fb6546709e8ec395496e82415fab714cab0ac6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR