Ethynyl-p-tolyl sulfone ≥98.0%
Supplier: TCI America
Synonyms:
p-Toluenesulfonylacetylene, Ethynyl p-Tolyl Sulfone, Tosylacetylene, Ethynyl p-Tolyl sulphone, p-Toluenesulphonylacetylene
E0466-5G
E0466-1G
TCE0466-5GEA
1009.62
CAD
TCE0466-5G
TCE0466-001G
Ethynyl-p-tolyl sulfone ≥98.0%
Ethynyl-p-tolyl sulfone
CAS Number: 13894-21-8
MDL Number: MFCD00191647
Molecular Formula: C9H8O2S
Molecular Weight: 180.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Sulfones, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: p-Toluenesulfonylacetylene, Tosylacetylene
Formula:
C₉H₈O₂S |
MDL Number:
MFCD00191647 CAS Number: 13894-21-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...