4,5-Ethylenedithio-1,3-dithiol-2-one ≥98.0%
Supplier: TCI America
E0460-5G
E0460-1G
TCE0460-005GEA
722.38
CAD
TCE0460-005G
TCE0460-001G
4,5-Ethylenedithio-1,3-dithiol-2-one ≥98.0%
4,5-Ethylenedithio-1,3-dithiol-2-one
CAS Number: 74962-29-1
MDL Number: MFCD00137883
Molecular Formula: C5H4OS4
Molecular Weight: 208.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 129
Formula:
C₅H₄OS₄ Melting Pt: 129 °C |
MDL Number:
MFCD00137883 CAS Number: 74962-29-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...