2-Amino-3-chloro-1,4-naphthoquinone ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

A1288-25G A1288-500G
TCA1288-025GEA 125.36 CAD
TCA1288-025G TCA1288-500G
2-Amino-3-chloro-1,4-naphthoquinone ≥98.0% (by HPLC, total nitrogen)
2-Amino-3-chloro-1,4-naphthoquinone

CAS Number: 2797-51-5
MDL Number: MFCD00001680
Molecular Formula: C10H6ClNO2
Molecular Weight: 207.61
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 200

Formula: C₁₀H₆ClNO₂
Melting Pt: 200 °C
MDL Number: MFCD00001680
CAS Number: 2797-51-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9039d47089e12836fb8bdfa588b93b1ce8027074&vid=9039d47089e12836fb8bdfa588b93b1ce8027074&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR