Ethyl (S)-(-)-1-ethyl-2-pyrrolidinecarboxylate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(-)-1-Ethyl-L-proline Ethyl Ester, (S)-(-)-1-Ethyl-2-pyrrolidinecarboxylic Acid Ethyl Ester, Et-Pro-OEt
CAS Number: 938-54-5
MDL Number: MFCD00191434
Molecular Formula: C9H17NO2
Molecular Weight: 171.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 0.97
Specific rotation [a]20/D: -94 deg (C=0.6, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: (S)-(-)-1-Ethyl-2-pyrrolidinecarboxylic Acid Ethyl Ester, (-)-1-Ethyl-L-proline Ethyl Ester, Et-Pro-OEt
Formula:
C₉H₁₇NO₂ MW: 171.24 g/mol Density: 0.97 (20 °C) |
MDL Number:
MFCD00191434 CAS Number: 938-54-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...