cis-Epoxysuccinic acid ≥97.0%
Supplier: TCI America
Synonyms:
cis-2,3-Oxiranedicarboxylic Acid
E0449-5G
E0449-25G
TCE0449-005GEA
160.73
CAD
TCE0449-005G
TCE0449-025G
cis-Epoxysuccinic acid ≥97.0%
cis-Epoxysuccinic acid
CAS Number: 16533-72-5
MDL Number: MFCD00191645
Molecular Formula: C4H4O5
Molecular Weight: 132.07
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 150
Other Product Names / Synonyms: cis-2,3-Oxiranedicarboxylic Acid
Formula:
C₄H₄O₅ MW: 132.07 g/mol Melting Pt: 150 °C |
MDL Number:
MFCD00191645 CAS Number: 16533-72-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...