2-Ethylthiophenothiazine ≥97.0% (by total nitrogen basis)

Supplier: TCI America

E0447-25G E0447-500G
TCE0447-025GEA 44.24 CAD
TCE0447-025G TCE0447-500G
2-Ethylthiophenothiazine ≥97.0% (by total nitrogen basis)
2-Ethylthiophenothiazine

CAS Number: 46815-10-5
MDL Number: MFCD00191362
Molecular Formula: C14H13NS2
Molecular Weight: 259.39
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 100

Formula: C₁₄H₁₃NS₂
Melting Pt: 100 °C
MDL Number: MFCD00191362
CAS Number: 46815-10-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0552b0f87524f0886a1a7a383af6a503e5fe56b6&vid=0552b0f87524f0886a1a7a383af6a503e5fe56b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR