Ethyl-2-ethyl-2-methylacetoacetate ≥93.0%

Supplier: TCI America

Synonyms: 2-Ethyl-2-methylacetoacetic Acid Ethyl Ester

E0437-25ML E0437-5ML
TCE0437-025MLEA 162.74 CAD
TCE0437-025ML TCE0437-5ML
Ethyl-2-ethyl-2-methylacetoacetate ≥93.0%
Ethyl-2-ethyl-2-methylacetoacetate

CAS Number: 33697-53-9
MDL Number: MFCD00191433
Molecular Formula: C9H16O3
Molecular Weight: 172.22
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 199
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: 2-Ethyl-2-methylacetoacetic Acid Ethyl Ester

Formula: C₉H₁₆O₃
MW: 172.22 g/mol
Boiling Pt: 199 °C
Density: 0.98 (20 °C)
MDL Number: MFCD00191433
CAS Number: 33697-53-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed68c0fbe0ed9b4631c7f8f19cdc71d3f0538b1f&vid=ed68c0fbe0ed9b4631c7f8f19cdc71d3f0538b1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR