Allyl-p-tolyl ether ≥95.0%

Supplier: TCI America

Synonyms: 4-Allyloxytoluene, Allyl p-Tolyl Ether, Allylp-tolylether

A1287-5G A1287-25G
TCA1287-005GEA 165.9 CAD
TCA1287-005G TCA1287-25G
Allyl-p-tolyl ether ≥95.0%
Allyl-p-tolyl ether

CAS Number: 23431-48-3
MDL Number: MFCD00026098
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.97


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4-Allyloxytoluene

Formula: C₁₀H₁₂O
MW: 148.21 g/mol
Density: 0.97 (20 °C)
MDL Number: MFCD00026098
CAS Number: 23431-48-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=003f964f52886336e4134e368ad25762a361b0fb&vid=003f964f52886336e4134e368ad25762a361b0fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR