EDTA trisodium salt hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Trisodium hydrogen ethylenediaminetetraacetate hydrate, Ethylenediaminetetra-acetic acid trisodium salt hydrate, Edetic acid trisodium salt hydrate, Ethylenedinitrilotetra-acetic acid trisodium salt hydrate
CAS Number: 85715-60-2
MDL Number: MFCD00013305
Molecular Formula: C10H16N2O8
Molecular Weight: 358.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Bioscience, Inhibitors (for Pharmacological Research)
Other Product Names / Synonyms: Ethylenediaminetetraacetic Acid Trisodium Salt, EDTA 3Na
Formula:
C₁₀H₁₃N₂Na₃O₈·H₂O MW: 376.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149675 CAS Number: 85715-60-2 EINECS: 205-758-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...