4,5-Ethylenedithio-1,3-dithiole-2-thione ≥98.0%

Supplier: TCI America

E0429-25G E0429-5G E0429-1G
TCE0429-025GEA 1638.46 CAD
TCE0429-025G TCE0429-005G TCE0429-1G
4,5-Ethylenedithio-1,3-dithiole-2-thione ≥98.0%
4,5-Ethylenedithio-1,3-dithiole-2-thione

CAS Number: 59089-89-3
MDL Number: MFCD00137882
Molecular Formula: C5H4S5
Molecular Weight: 224.39
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 124

Formula: C₅H₄S₅
MW: 224.39 g/mol
Melting Pt: 124 °C
MDL Number: MFCD00137882
CAS Number: 59089-89-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13fcb0da73e250c9cf16c5d86378dddb72226de9&vid=13fcb0da73e250c9cf16c5d86378dddb72226de9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR