(4-Ethoxybenzyl)triphenylphosphonium bromide ≥98.0% for synthesis

Supplier: TCI America

E0421-5G E0426-10G
TCE0421-5GEA 80.58 CAD
TCE0421-5G TCE0426-010G
(4-Ethoxybenzyl)triphenylphosphonium bromide ≥98.0% for synthesis
(4-Ethoxybenzyl)triphenylphosphonium bromide

CAS Number: 622-38-8
MDL Number: MFCD00009265
Molecular Formula: C8H10S
Molecular Weight: 138.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 71
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: Ethylthiobenzene

Formula: C₂₇H₂₆BrOP
MW: 477.370 g/mol
Boiling Pt: 205 °C
Melting Pt: 225 °C
Density: 1.02 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00011834
CAS Number: 82105-88-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fd6f68a84eac7b62bc864def4bc063f81cef90e&vid=0fd6f68a84eac7b62bc864def4bc063f81cef90e&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f743ceca2b14609992a3d7ae319abb576e20c4c4&vid=f743ceca2b14609992a3d7ae319abb576e20c4c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR