N-Ethyl-4-chloroaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0420-25G
TCE0420-025GEA 125.61 CAD
TCE0420-025G
N-Ethyl-4-chloroaniline ≥98.0% (by GC, titration analysis)
N-Ethyl-4-chloroaniline

CAS Number: 13519-75-0
MDL Number: MFCD00018594
Molecular Formula: C8H10ClN
Molecular Weight: 155.63
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 101
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 4-Chloro-N-ethylaniline

Formula: C₈H₁₀ClN
Boiling Pt: 101 °C
Density: 1.12 (20 °C)
MDL Number: MFCD00018594
CAS Number: 13519-75-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03bd09f545295467540de7badc7247805745dd1c&vid=03bd09f545295467540de7badc7247805745dd1c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR