Ethylene glycolbis(4-carboxyphenyl)ether ≥95.0%

Supplier: TCI America

Synonyms: 4,4'-(Ethane-1,2-diylbis(oxy))dibenzoic acid, 1,2-Bis(4-carboxyphenoxy)ethane

E0415-1G E0415-5G
TCE0415-1GEA 331.8 CAD
TCE0415-1G TCE0415-005G
Ethylene glycolbis(4-carboxyphenyl)ether ≥95.0%
Ethylene glycolbis(4-carboxyphenyl)ether

CAS Number: 3753-05-7
MDL Number: MFCD00020365
Molecular Formula: C16H14O6
Molecular Weight: 302.28
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Bis(4-carboxyphenoxy)ethane

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00020365
CAS Number: 3753-05-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7447a14d99636b28323bf504d111573dfc9a5eb&vid=d7447a14d99636b28323bf504d111573dfc9a5eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR