Ethyl-2-ethoxybenzoate ≥98.0%

Supplier: TCI America

Synonyms: 2-Ethoxybenzoic Acid Ethyl Ester, Ethyl 2-ethoxybenzoate

E0413-25G E0413-500G
TCE0413-025GEA 146.94 CAD
TCE0413-025G TCE0413-500G
Ethyl-2-ethoxybenzoate ≥98.0%
Ethyl-2-ethoxybenzoate

CAS Number: 6290-24-0
MDL Number: MFCD00142907
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 251
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: 2-Ethoxybenzoic Acid Ethyl Ester

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 251 °C
Density: 1.07 (20 °C)
MDL Number: MFCD00142907
CAS Number: 6290-24-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b96ecf98faaf46239cfcb8e5d83857c641c107c2&vid=b96ecf98faaf46239cfcb8e5d83857c641c107c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR