2-Ethyl-2-methyl-1,3-propanediol ≥97.0%

Supplier: TCI America

Synonyms: 2,2-Bis(hydroxymethyl)butane

E0412-5G E0412-25G
TCE0412-5GEA 284.24 CAD
TCE0412-5G TCE0412-025G
2-Ethyl-2-methyl-1,3-propanediol ≥97.0%
2-Ethyl-2-methyl-1,3-propanediol

CAS Number: 77-84-9
MDL Number: MFCD00004693
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 226
Melting point (°C): 43
Flash Point (°C): 110


Other Product Names / Synonyms: 2,2-Bis(hydroxymethyl)butane

Formula: C₆H₁₄O₂
Boiling Pt: 226 °C
Melting Pt: 43 °C
Flash Pt: 110 °C
MDL Number: MFCD00004693
CAS Number: 77-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e800e9a31f205774ae2b735c2713bdda7219c42f&vid=e800e9a31f205774ae2b735c2713bdda7219c42f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR