Vinyl-2-ethylhexanoate ≥98.0% stabilized
Supplier: TCI America
E0404-25ML
E0404-500ML
TCE0404-025MLEA
41.08
CAD
TCE0404-025ML
TCE0404-500ML
Vinyl-2-ethylhexanoate ≥98.0% stabilized
Vinyl-2-ethylhexanoate
(stabilized with MEHQ)
CAS Number: 94-04-2
MDL Number: MFCD00009488
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 65
Specific Gravity (20/20): 0.87
Other Product Names / Synonyms: 2-Ethylhexanoic Acid Vinyl Ester
Formula:
C₁₀H₁₈O₂ Boiling Pt: 86 °C Density: 0.87 (20 °C) Flash Pt: 65 °C |
MDL Number:
MFCD00009488 CAS Number: 94-04-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...