N,N,N',N'-Ethylenediaminetetrakis(methylenephosphonic acid) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: ((Ethane-1,2-diylbis(azanetriyl))tetrakis(methylene))tetraphosphonic acid

E0393-25G E0393-500G E0393-100G
TCE0393-025GEA 92.59 CAD
TCE0393-025G TCE0393-500G TCE0393-100G
N,N,N',N'-Ethylenediaminetetrakis(methylenephosphonic acid) ≥98.0% (by titrimetric analysis)
N,N,N',N'-Ethylenediaminetetrakis(methylenephosphonic acid)

CAS Number: 1429-50-1
MDL Number: MFCD00058893
Molecular Formula: C6H20N2O12P4
Molecular Weight: 436.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₆H₂₀N₂O₁₂P₄
MW: 436.12 g/mol
MDL Number: MFCD00058893
CAS Number: 1429-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=889612953335ed3fd6a37bb427c5027027825fe8&vid=889612953335ed3fd6a37bb427c5027027825fe8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR