3-Ethoxyacrylonitrile (cis- and trans- mixture) ≥98.0%

Supplier: TCI America

E0388-250ML E0388-25ML
TCE0388-250MLEA 758.4 CAD
TCE0388-250ML TCE0388-025ML
3-Ethoxyacrylonitrile (cis- and trans- mixture) ≥98.0%
3-Ethoxyacrylonitrile (cis- and trans- mixture)

CAS Number: 61310-53-0
MDL Number: MFCD00010193
Molecular Formula: C5H7NO
Molecular Weight: 97.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 71
Flash Point (°C): 82
Specific Gravity (20/20): 0.95

Formula: C₅H₇NO
Boiling Pt: 71 °C
Density: 0.95 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00010193
CAS Number: 61310-53-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d804dbd0d65b08a18b03a01605430b43ef49c7f7&vid=d804dbd0d65b08a18b03a01605430b43ef49c7f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR