Ethyl-2-chloro-2-(hydroxyimino)acetate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Chloro-2-(hydroxyimino)acetic acid ethyl ester, alpha-Chloroglyoxalic acid ethyl ester Oxime, Ethylchloroximidoacetate, Ethyl 2-Chloro-2-(hydroxyimino)acetate

E0371-5G E0371-25G
TCE0371-005GEA 66.36 CAD
TCE0371-005G TCE0371-25G
Ethyl-2-chloro-2-(hydroxyimino)acetate ≥99.0% (by titrimetric analysis)
Ethyl-2-chloro-2-(hydroxyimino)acetate

CAS Number: 14337-43-0
MDL Number: MFCD00010209
Molecular Formula: C4H6ClNO3
Molecular Weight: 151.55
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 81


Other Product Names / Synonyms: alpha-Chloroglyoxalic Acid Ethyl Ester Oxime, 2-Chloro-2-(hydroxyimino)acetic Acid Ethyl Ester, Ethylchloroximidoacetate

Formula: C₄H₆ClNO₃
MW: 151.5483 g/mol
Melting Pt: 81 °C
MDL Number: MFCD00010209
CAS Number: 14337-43-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=386eb4b84a5ab03c1b41a78ca93e00e91a49f6e8&vid=386eb4b84a5ab03c1b41a78ca93e00e91a49f6e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR