1-Ethyl-4-(methoxycarbonyl)pyridinium iodide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Carbomethoxy-1-ethylpyridinium Iodide

E0362-1G E0362-5G
TCE0362-001GEA 42.66 CAD
TCE0362-001G TCE0362-005G
1-Ethyl-4-(methoxycarbonyl)pyridinium iodide ≥97.0% (by titrimetric analysis)
1-Ethyl-4-(methoxycarbonyl)pyridinium iodide

CAS Number: 1199-65-1
MDL Number: MFCD00011996
Molecular Formula: C9H12INO2
Molecular Weight: 293.10
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 110


Other Product Names / Synonyms: 4-Carbomethoxy-1-ethylpyridinium Iodide

Formula: C₉H₁₂INO₂
MW: 293.10 g/mol
Melting Pt: 110 °C
MDL Number: MFCD00011996
CAS Number: 1199-65-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=075b5501805459454c90cad102d0356f879846bd&vid=075b5501805459454c90cad102d0356f879846bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR