N-Ethylcarbazole-3-carboxaldehyde ≥98.0%

Supplier: TCI America

Synonyms: N-Ethyl-3-carbazolecarboxaldehyde, 3-Formyl-N-ethylcarbazole

E0359-25G E0359-5G
TCE0359-025GEA 299.25 CAD
TCE0359-025G TCE0359-5G
N-Ethylcarbazole-3-carboxaldehyde ≥98.0%
N-Ethylcarbazole-3-carboxaldehyde

CAS Number: 7570-45-8
MDL Number: MFCD00004963
Molecular Formula: C15H13NO
Molecular Weight: 223.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 255
Melting point (°C): 90


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-Formyl-N-ethylcarbazole

Formula: C₁₅H₁₃NO
Boiling Pt: 255 °C
Melting Pt: 90 °C
MDL Number: MFCD00004963
CAS Number: 7570-45-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=534291dae2d7301eed524be2d73a604a6a1327f0&vid=534291dae2d7301eed524be2d73a604a6a1327f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR