N-Ethoxycarbonyl-5-nitro-o-toluidine ≥96.0%
Supplier: TCI America
Synonyms:
N-(5-Nitro-2-methylphenyl)urethan, Ethyl 2-Methyl-5-nitrophenylcarbamate, N-Carbethoxy-5-nitro-o-toluidine
E0349-25G
E0349-5G
TCE0349-025GEA
239.69
CAD
TCE0349-025G
TCE0349-5G
N-Ethoxycarbonyl-5-nitro-o-toluidine ≥96.0%
N-Ethoxycarbonyl-5-nitro-o-toluidine
CAS Number: 16648-52-5
MDL Number: MFCD00059827
Molecular Formula: C10H12N2O4
Molecular Weight: 224.22
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 135
Other Product Names / Synonyms: N-Carbethoxy-5-nitro-o-toluidine, Ethyl 2-Methyl-5-nitrophenylcarbamate, N-(5-Nitro-2-methylphenyl)urethan
Formula:
C₁₀H₁₂N₂O₄ MW: 224.22 g/mol Melting Pt: 135 °C |
MDL Number:
MFCD00059827 CAS Number: 16648-52-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...