4,4'-Ethylenedianiline ≥97.0%
Supplier: TCI America
Synonyms:
4,4'-Diamino-1,2-diphenylethane, 4,4'-Diaminobibenzyl, alpha,alpha'-Bi-p-toluidine, 4,4'-(Ethane-1,2-diyl)dianiline
CAS Number: 621-95-4
MDL Number: MFCD00007923
Molecular Formula: C14H16N2
Molecular Weight: 212.30
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 134
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: alpha,alpha'-Bi-p-toluidine, 4,4'-Diaminobibenzyl, 4,4'-Diamino-1,2-diphenylethane
Formula:
C₁₄H₁₆N₂ MW: 212.30 g/mol Melting Pt: 134 °C |
MDL Number:
MFCD00007923 CAS Number: 621-95-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...