Ethylene glycol diglycidyl ether (so called)

Supplier: TCI America

Synonyms: Quetol 651, 1,2-Diglycidyloxyethane, 1,2-Bis(2,3-epoxypropoxy)ethane

E0342-25G E0342-500G
TCE0342-025GEA 65.38 CAD
TCE0342-025G TCE0342-500G
Ethylene glycol diglycidyl ether (so called)
Ethylene glycol diglycidyl ether

CAS Number: 2224-15-9
MDL Number: MFCD00005142
Molecular Formula: C8H14O4
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 157
Specific Gravity (20/20): 1.19


Other Product Names / Synonyms: 1,2-Bis(2,3-epoxypropoxy)ethane, 1,2-Diglycidyloxyethane

Formula: C₈H₁₄O₄
MW: 174.19 g/mol
Density: 1.19 (20 °C)
Flash Pt: 157 °C
MDL Number: MFCD00005142
CAS Number: 2224-15-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c66ed435be7cb339b107b738c4fc8777b2247a0&vid=8c66ed435be7cb339b107b738c4fc8777b2247a0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR