3-Aminophenylboronic acid monohydrate (contains varying amounts of Anhydride) for synthesis

Supplier: TCI America

Synonyms: Dihydroxy(3-aminophenyl)borane monohydrate, 3-Aminobenzeneboronic acid monohydrate, 3-Aminobenzeneboronic acid, 3-Aminophenylboronic Acid Monohydrate, (3-Aminophenyl)metaboric acid monohydrate

A1281-5G A1281-25G A1281-1G
TCA1281-005GEA 254.38 CAD
TCA1281-005G TCA1281-25G TCA1281-001G
3-Aminophenylboronic acid monohydrate (contains varying amounts of Anhydride) for synthesis
3-Aminophenylboronic acid monohydrate

CAS Number: 206658-89-1
MDL Number: MFCD00007755
Molecular Formula: C6H8BNO2
Molecular Weight: 136.95
Form: Crystal
Color: Very Pale Yellow


Other Product Names / Synonyms: 3-Aminobenzeneboronic Acid

Formula: C₆H₈BNO₂·1H₂O
MW: 154.96 g/mol
MDL Number: MFCD00149554
CAS Number: 206658-89-1
EINECS: 250-189-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80c6cd4c5480ae5412383e81fd50dfa6213605ff&vid=80c6cd4c5480ae5412383e81fd50dfa6213605ff&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR