Dimethyl Icosanedioate ≥95.0%

Supplier: TCI America

E0330-5G E0330-25G
TCE0330-005GEA 204.93 CAD
TCE0330-005G TCE0330-025G
Dimethyl Icosanedioate ≥95.0%
Dimethyl Icosanedioate

CAS Number: 42235-38-1
MDL Number: MFCD00059302
Molecular Formula: C22H42O4
Molecular Weight: 370.57
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 223
Melting point (°C): 66


Other Product Names / Synonyms: Dimethyl Eicosanedioate, Eicosanedioic Acid Dimethyl Ester, Icosanedioic Acid Dimethyl Ester

Formula: C₂₂H₄₂O₄
Boiling Pt: 223 °C
Melting Pt: 66 °C
MDL Number: MFCD00059302
CAS Number: 42235-38-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bab1729ca94106f23bcfbb6b0a40047c3ee2a95&vid=5bab1729ca94106f23bcfbb6b0a40047c3ee2a95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR