2-(3,4-Epoxycyclohexyl)ethyltrimethoxysilane ≥97.0%
Supplier: TCI America
Synonyms:
Trimethoxy[2-(7-oxabicyclo[4.1.0]hept-3-yl)ethyl]silane, Trimethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane
CAS Number: 3388-04-3
MDL Number: MFCD00014485
Molecular Formula: C11H22O4Si
Molecular Weight: 246.38
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 310
Flash Point (°C): 141
Specific Gravity (20/20): 1.07
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Trimethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane
Formula:
C₁₁H₂₂O₄Si MW: 246.38 g/mol Boiling Pt: 310 °C Density: 1.07 (20 °C) Flash Pt: 141 °C |
MDL Number:
MFCD00014485 CAS Number: 3388-04-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...