Ethylene trithiocarbonate ≥96.0%
Supplier: TCI America
E0323-25G
E0323-100G
TCE0323-025GEA
262.06
CAD
TCE0323-025G
TCE0323-100G
Ethylene trithiocarbonate ≥96.0%
Ethylene trithiocarbonate
CAS Number: 822-38-8
MDL Number: MFCD00005493
Molecular Formula: C3H4S3
Molecular Weight: 136.25
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Flash Point (°C): 163
Freezing point (°C): 33
Other Product Names / Synonyms: 1,3-Dithiolane-2-thione
Formula:
C₃H₄S₃ Flash Pt: 163 °C |
MDL Number:
MFCD00005493 CAS Number: 822-38-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...