Ethylene glycol mono-p-tolyl ether ≥98.0%
Supplier: TCI America
Synonyms:
2-(p-Tolyloxy)ethanol, 4-(2-Hydroxyethoxy)toluene, 2-(4-Methylphenoxy)ethanol
E0303-5G
E0303-25G
TCE0303-005GEA
124.19
CAD
TCE0303-005G
TCE0303-025G
Ethylene glycol mono-p-tolyl ether ≥98.0%
Ethylene glycol mono-p-tolyl ether
CAS Number: 15149-10-7
MDL Number: MFCD00020600
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 123
Melting point (°C): 45
Other Product Names / Synonyms: 4-(2-Hydroxyethoxy)toluene, 2-(4-Methylphenoxy)ethanol
Formula:
C₉H₁₂O₂ MW: 152.19 g/mol Boiling Pt: 123 °C Melting Pt: 45 °C |
MDL Number:
MFCD00020600 CAS Number: 15149-10-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...