1,2-Bis(carboxymethylthio)ethane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0278-25G E0278-500G E0278-100G
TCE0278-025GEA 157.05 CAD
TCE0278-025G TCE0278-500G TCE0278-100G
1,2-Bis(carboxymethylthio)ethane ≥98.0% (by GC, titration analysis)
1,2-Bis(carboxymethylthio)ethane

CAS Number: 7244-02-2
MDL Number: MFCD00021771
Molecular Formula: C6H10O4S2
Molecular Weight: 210.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 110


Other Product Names / Synonyms: 1,2-Bis(carboxymethylthio)ethane, 1,2-Ethylenebis(thioglycolic Acid)

Formula: C₆H₁₀O₄S₂
Melting Pt: 110 °C
MDL Number: MFCD00021771
CAS Number: 7244-02-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc9d750cbaa0a464671e0e21a7607accdd2b1456&vid=fc9d750cbaa0a464671e0e21a7607accdd2b1456&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR