Ethylenediamine-N,N'-diacetic acid ≥98.0%

Supplier: TCI America

Synonyms: Ethylene-N,N'-diglycine, EDDA

E0275-5G E0275-25G
TCE0275-5GEA 139.04 CAD
TCE0275-5G TCE0275-025G
Ethylenediamine-N,N'-diacetic acid ≥98.0%
Ethylenediamine-N,N'-diacetic acid

CAS Number: 5657-17-0
MDL Number: MFCD00004281
Molecular Formula: C6H12N2O4
Molecular Weight: 176.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 200


Other Product Names / Synonyms: Ethylene-N,N'-diglycine, EDDA

Formula: C₆H₁₂N₂O₄
MW: 176.17 g/mol
Melting Pt: 200 °C
MDL Number: MFCD00004281
CAS Number: 5657-17-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df38fa639bc19d672306ffda288d303cf78c0c16&vid=df38fa639bc19d672306ffda288d303cf78c0c16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR