4-Ethyl-1-octyn-3-ol ≥97.0%

Supplier: TCI America

E0270-25ML E0270-500ML
TCE0270-25MLEA 87.53 CAD
TCE0270-25ML TCE0270-500ML
4-Ethyl-1-octyn-3-ol ≥97.0%
4-Ethyl-1-octyn-3-ol

CAS Number: 5877-42-9
MDL Number: MFCD00015262
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Specific Gravity (20/20): 0.87


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: (2-Ethyl-1-hydroxyhexyl)acetylene

Formula: C₁₀H₁₈O
Boiling Pt: 126 °C
Density: 0.87 (20 °C)
MDL Number: MFCD00015262
CAS Number: 5877-42-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dba1531791688d1ebc63d1739ef9a6569b4dfeba&vid=dba1531791688d1ebc63d1739ef9a6569b4dfeba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR