4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers) ≥95.0% (by GC, titration analysis)

Supplier: TCI America

A1278-25G A1278-5G
TCA1278-025GEA 380.78 CAD
TCA1278-025G TCA1278-005G
4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers) ≥95.0% (by GC, titration analysis)
4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers)

CAS Number: 1776-53-0
MDL Number: MFCD00191601
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Aminohexahydrobenzoic Acid

Formula: C₇H₁₃NO₂
MDL Number: MFCD00191601
CAS Number: 1776-53-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=174bc572b634c1adb090a79c1902b5356ebd2368&vid=229cdc212949f86f15efafe9e2ccfa4709652539&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR