N-(4-Ethoxybenzylidene)-4-butylaniline ≥99.0% (by titrimetric analysis)

Supplier: TCI America

E0262-25G E0262-500G
TCE0262-025GEA 167.13 CAD
TCE0262-025G TCE0262-500G
N-(4-Ethoxybenzylidene)-4-butylaniline ≥99.0% (by titrimetric analysis)
N-(4-Ethoxybenzylidene)-4-butylaniline

CAS Number: 29743-08-6
MDL Number: MFCD00009465
Molecular Formula: C19H23NO
Molecular Weight: 281.40
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Flash Point (°C): 113
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: EBBA

Formula: C₁₉H₂₃NO
Flash Pt: 113 °C
MDL Number: MFCD00009465
CAS Number: 29743-08-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=956272547737181dd0506f52522cfd9aa0147688&vid=956272547737181dd0506f52522cfd9aa0147688&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR