2-Ethylhexyl glycidyl ether ≥98.0%

Supplier: TCI America

E0229-25ML E0229-500ML
TCE0229-025MLEA 55.3 CAD
TCE0229-025ML TCE0229-500ML
2-Ethylhexyl glycidyl ether ≥98.0%
2-Ethylhexyl glycidyl ether

CAS Number: 2461-15-6
MDL Number: MFCD00005141
Molecular Formula: C11H22O2
Molecular Weight: 186.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 61
Flash Point (°C): 94
Specific Gravity (20/20): 0.90


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: (2-Ethylhexyloxy)-2,3-epoxypropane

Formula: C₁₁H₂₂O₂
Boiling Pt: 61 °C
Density: 0.90 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00005141
CAS Number: 2461-15-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR