2-Amino-p-cresol hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 2-Hydroxy-5-methylaniline Hydrochloride, 2-Amino-4-methylphenol Hydrochloride

A1276-25G A1276-500G
TCA1276-025GEA 45.82 CAD
TCA1276-025G TCA1276-500G
2-Amino-p-cresol hydrochloride ≥98.0%
2-Amino-p-cresol hydrochloride

CAS Number: 2977-71-1
MDL Number: MFCD00191600
Molecular Formula: C7H9NO
Molecular Weight: 159.61
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2-Amino-4-methylphenol Hydrochloride, 2-Hydroxy-5-methylaniline Hydrochloride

Formula: C₇H₉NO
MW: 159.61 g/mol
MDL Number: MFCD00191600
CAS Number: 2977-71-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc7fd20d5d428ce81a7f08acdb285554e3cf7db&vid=dcc7fd20d5d428ce81a7f08acdb285554e3cf7db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR