Ethylene glycol dibutyl ether ≥98.0%

Supplier: TCI America

E0217-25ML E0217-500ML
TCE0217-025MLEA 88.4 CAD
TCE0217-025ML TCE0217-500ML
Ethylene glycol dibutyl ether ≥98.0%
Ethylene glycol dibutyl ether

CAS Number: 112-48-1
MDL Number: MFCD00048785
Molecular Formula: C10H22O2
Molecular Weight: 174.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 202
Melting point (°C): -69
Flash Point (°C): 85
Specific Gravity (20/20): 0.84


Other Product Names / Synonyms: 1,2-Dibutoxyethane, Dibutyl Cellosolve, Dibutyl Glycol

Formula: C₁₀H₂₂O₂
Boiling Pt: 202 °C
Melting Pt: –69 °C
Density: 0.84 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00048785
CAS Number: 112-48-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e5c248071adc19c51e4fc708918eff1c4d8525&vid=a0e5c248071adc19c51e4fc708918eff1c4d8525&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR