Sodium ethylxanthate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Sodium Xanthogenate, Ethylxanthic Acid Sodium Salt, Sodium O-Ethyl Dithiocarbonate

E0195-500G E0195-25G
TCE0195-500GEA 1300.34 CAD
TCE0195-500G TCE0195-025G
Sodium ethylxanthate ≥97.0% (by titrimetric analysis)
Sodium ethylxanthate

CAS Number: 140-90-9
MDL Number: MFCD00068314
Molecular Formula: C3H6OS2
Molecular Weight: 144.18
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Other Product Names / Synonyms: Ethylxanthic Acid Sodium Salt, Sodium O-Ethyl Dithiocarbonate, Sodium Xanthogenate

Formula: C₃H₆OS₂
MW: 144.18 g/mol
MDL Number: MFCD00068314
CAS Number: 140-90-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5da7b0dd62d10a9b60a822489082b8fabbb0a406&vid=5da7b0dd62d10a9b60a822489082b8fabbb0a406&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR