4-Amino-2,6-diphenylphenol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A1269-1G A1269-500MG
TCA1269-001GEA 135.88 CAD
TCA1269-001G TCA1269-500MG
4-Amino-2,6-diphenylphenol ≥98.0% (by titrimetric analysis)
4-Amino-2,6-diphenylphenol

CAS Number: 50432-01-4
MDL Number: MFCD00034070
Molecular Formula: C18H15NO
Molecular Weight: 261.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 148


Other Product Names / Synonyms: 5'-Amino-2'-hydroxy-m-terphenyl

Formula: C₁₈H₁₅NO
Melting Pt: 148 °C
MDL Number: MFCD00034070
CAS Number: 50432-01-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33d2c8560cc2ab4fa63e955a3811f48bdb68e5c2&vid=33d2c8560cc2ab4fa63e955a3811f48bdb68e5c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR