Ethyl phthalyl ethyl glycolate ≥95.0%

Supplier: TCI America

Synonyms: Ethoxycarbonylmethyl Ethyl Phthalate, Phthalic Acid Ethoxycarbonylmethyl Ethyl Ester

E0164-25G E0164-500G
TCE0164-025GEA 26.86 CAD
TCE0164-025G TCE0164-500G
Ethyl phthalyl ethyl glycolate ≥95.0%
Ethyl phthalyl ethyl glycolate

CAS Number: 84-72-0
MDL Number: MFCD00059377
Molecular Formula: C14H16O6
Molecular Weight: 280.28
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 320
Flash Point (°C): 185
Freezing point (°C): 13
Specific Gravity (20/20): 1.19


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Ethoxycarbonylmethyl Ethyl Phthalate, Phthalic Acid Ethoxycarbonylmethyl Ethyl Ester

Formula: C₁₄H₁₆O₆
MW: 280.28 g/mol
Boiling Pt: 320 °C
Density: 1.19 (20 °C)
Flash Pt: 185 °C
MDL Number: MFCD00059377
CAS Number: 84-72-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2328fb67eb9eeea0ebeee5026ff0cdc4b6a9eaca&vid=2328fb67eb9eeea0ebeee5026ff0cdc4b6a9eaca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR