Zinc-N-ethyl-N-phenyldithiocarbamate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
E0163-25G
E0163-500G
TCE0163-025GEA
44.24
CAD
TCE0163-025G
TCE0163-500G
Zinc-N-ethyl-N-phenyldithiocarbamate ≥98.0% (by titrimetric analysis)
Zinc-N-ethyl-N-phenyldithiocarbamate
CAS Number: 14634-93-6
MDL Number: MFCD00059686
Molecular Formula: C9H11NS2
Molecular Weight: 457.99
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 206
Flash Point (°C): 209
Other Product Names / Synonyms: N-Ethyl-N-phenyldithiocarbamic Acid Zinc Salt
Formula:
C₉H₁₁NS₂ Melting Pt: 206 °C Flash Pt: 209 °C |
MDL Number:
MFCD00059686 CAS Number: 14634-93-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...