N-Ethyl-2-naphthylamine hydrobromide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0156-5G E0156-25G
TCE0156-5GEA 117.39 CAD
TCE0156-5G TCE0156-025G
N-Ethyl-2-naphthylamine hydrobromide ≥98.0% (by GC, titration analysis)
N-Ethyl-2-naphthylamine hydrobromide

MDL Number: MFCD00060158
Molecular Formula: C12H13N
Molecular Weight: 252.16
Purity/Analysis Method: >98.0% (T,GC)
Form: Crystal

Formula: C₁₂H₁₃N.HBr
MDL Number: MFCD00060158
CAS Number: 381670-27-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e840f3a884273ea1b730b84cf7a0b45411b4bdb&vid=4e840f3a884273ea1b730b84cf7a0b45411b4bdb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T,GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR